Air Compressor, Precision, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
65-1555
103549-794EA 8410.18 USD
103549-794
Air Compressor, Precision, Peak Scientific
Gas Generators Compressed Air Stations
The Precision Air Compressor is designed and engineered specifically to provide a dedicated, fully integrated compressed air source for Precision Nitrogen and Precision Zero Air gas generators.
  • Compressed air supply for Precision Nitrogen and Zero Air generators
  • Single unit can supply both the Precision Nitrogen and Zero Air generators in a stack
  • Duplex compressor mounting system to dampen noise and vibration
  • Service and repair status indicators
  • 12 month comprehensive on-site warranty
The Precision Air Compressor is designed and engineeredMore Product Information