Zero Air Generator, Precision Zero Air 30 L, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
60-0031
103549-792EA 16398.05 USD
103549-792
Zero Air Generator, Precision Zero Air 30 L, Peak Scientific
Gas Generators Zero Air Generators
The Precision Zero Air 30L is engineered specifically to provide flame support gas to multiple GC instruments.
  • Integrates with existing Precision products, with same modular, stackable design
  • Hydrocarbon content (as Methane) < 0.05ppm for market-leading purity
  • Up to 30L/min of Zero Air Up to 100psi/6.9bar outlet pressure
  • Minimum maintenance requirements, no expensive catalyst chamber replacements
  • Greatly reduced risk of contaminants entering system and no risk of running out of gas
  • Stylish and contemporary aesthetics and functional design
  • Status indicative lighting feature (amber warmup, green ready)
  • Compatible with Precision Air Compressor
  • 12 month comprehensive on-site warranty
The Precision Zero Air 30L is engineered specifically to provide flame support gas to multiple GC instruments. It provides up to 30 litres per minute of hydrocarbon free air, which is suitable to support your FID, FPD and NPD at both standard and trace detection limits.
Expanding Peak Scientific’s Precision range, the Precision Zero Air 30L, has the same stackable form-factor and compact footprint found in the rest of the Precision Series, ensuring it integrates seamlessly with other Precision models to provide a convenient, reliable and safe gas solution for specific GC gas requirements.