Zero Air Generator, Precision Zero Air 7 L, Peak Scientfic
Supplier: PEAK SCIENTIFIC MS
60-0008
103549-788EA 9729.22 USD
103549-788
Zero Air Generator, Precision Zero Air 7 L, Peak Scientfic
Gas Generators Zero Air Generators
Precision Zero Air 7L is engineered specifically to provide flame support gas to multiple GC instruments.
  • Integrates with existing Precision products, with same modular, stackable design
  • Hydrocarbon content (as Methane) < 0.05ppm for market-leading purity
  • Up to 7 L/min of Zero Air
  • Up to 80 psi/5.5 bar outlet pressure
  • Minimum maintenance requirements, no expensive catalyst chamber replacements
  • Greatly reduced risk of contaminants entering system and no risk of running out of gas
  • Stylish and contemporary aesthetics and functional design
  • Status indicative lighting feature (amber warmup, green ready)
  • Compatible with Precision Air Compressor
  • 12 month comprehensive on-site warranty
Precision Zero Air 7 L is engineered specifically to provide flame support gas to multiple GC instruments. It provides up to 7 liters per minute of hydrocarbon free air, which is ideal for laboratories with multiple GC Detectors, such as FID, FPD, and NPD. The Precision Zero Air 7 L expands Peak Scientific's Precision range, with the same stackable form-factor and compact footprint ensuring it integrates seamlessly with other Precision modules to provide a convenient, reliable and safe gas solution for specific GC gas requirements.