Zero Air Generator, Precision Zero Air 3.5 L, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
60-3501
103549-786EA 7579.09 USD
103549-786
Zero Air Generator, Precision Zero Air 3.5 L, Peak Scientific
Gas Generators Zero Air Generators
The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC.
  • Integrates seamlessly with other Precision units
  • Hydrocarbon content (as Methane) < 0.05ppm for market leading purity
  • Minimum lifetime maintenance requirements, no expensive catalyst chamber replacements
  • Avoid risk of contaminants entering the system (when switching out empty cylinders)
  • No risk of running out of gas in middle of analysis
  • Compatible with Precision Air
  • Compressor Status indicative lighting feature (amber warm-up, green ready)
  • 12 month comprehensive on-site warranty
The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC. As with all Precision Series generators, Zero Air benefits from a compact and modular, stackable design, minimizing the total footprint required for GC gas supply, and providing flexibility to add or remove modules as your laboratory requirements evolve over time.