Zero Air Generator, Precision Zero Air 1.5, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
60-1501
103549-784EA 5637.69 USD
103549-784
Zero Air Generator, Precision Zero Air 1.5, Peak Scientific
Gas Generators Zero Air Generators
The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC at both standard and trace detection limits.
  • Integrates seamlessly with other Precision units Hydrocarbon content (as Methane) < 0.05ppm for market leading purity
  • Minimum lifetime maintenance requirements, no expensive catalyst chamber replacements
  • Avoid risk of contaminants entering the system (when switching out empty cylinders)
  • No risk of running out of gas in middle of analysis
  • Compatible with Precision Air Compressor
  • Status indicative lighting feature (amber warm-up, green ready)
  • 12 month comprehensive on-site warranty
The Precision Zero Air generators are designed specifically to supply clean, dry, hydrocarbon-free air to be used as flame support gas for GC at both standard and trace detection limits.
As with all Precision series generators, Zero Air benefits from a compact and modular, stackable design, minimizing the total footprint required for GC gas supply, and providing flexibility to add or remove modules as your laboratory requirements evolve over time.