Hydrogen Generator, Precision Hydrogen 300, Peak Scientific
Supplier: PEAK SCIENTIFIC MS
63-0300
103549-772EA 14260.88 USD
103549-772
Hydrogen Generator, Precision Hydrogen 300, Peak Scientific
Gas Generators Hydrogen Generators
The Precision Hydrogen generators are designed to provide the gas needed for detectors requiring hydrogen fuel gas, such as FID.
  • 99.9995% purity
  • Suitable for GC detector gas
  • Creates hydrogen on demand, minimal storage of hydrogen in the system
  • Low maintenance and minimal running costs over product lifetime
  • Internal leak detection with automatic shutdown
  • Automatic loading pump as standard
  • Simple maintenance, limited to replacing de-ionizer cartridge and silica gel
The Precision Hydrogen generators are designed to provide the gas needed for detectors requiring hydrogen fuel gas, such as FID. One generator is capable of supplying multiple detectors, and are available in various flow rates to suit individual customer needs.
These generators utilize a Proton Exchange Membrane (PEM) to create the hydrogen gas from deionized water and a desiccant filtration stage is used to dry the gas. The Precision Hydrogen generators are also suitable for providing collision gas for ICP-MS.
Precision Hydrogen gas generators come with various safety features as standard, giving you complete peace of mind in the laboratory and are a far safer, dependable and convenient alternative to cylinders.