BCA Protein Assay kit (BCA-PAK), OZ Biosciences
Supplier: OZ Biosciences
BCA2500
103549-592EA 203.08 USD
103549-592
BCA Protein Assay kit (BCA-PAK), OZ Biosciences
Assays Protein Assays
BCA Protein Assay kit (BCA-PAK) is useful for colorimetric detection and quantification of total protein content even in the presence of detergents.
This kit is based on the reduction of Cu2+ to Cu1+ by protein in alkaline solution; monovalent copper ions produced are detected in a concentration-dependent manner. Bicinchoninic acid (BCA) chelates with the reduced copper Cu1+ and forms a water-soluble purple reaction complex that exhibits a strong absorbance at 562 nm. Absorbance is linear over a wide range of protein concentrations between 25-2000 µg/mL. In general, protein concentrations are estimated with reference to a commonly used protein standard; the kit also includes Bovine Serum Albumin (BSA) at 1.5 mg/mL as a protein standard for a convenient preparation of protein concentration standard curves.      
Delivery information: Shipped at room temperature.
Packaging: Store at 4°C