RMESFect™ CRISPR Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
RMC70500
103537-334EA 264.62 USD
103537-334
RMESFect™ CRISPR Transfection Reagent, OZ Biosciences
Transfection Reagents
RmesFect™ CRISPR transfection Reagent based on the TEE-technology is specifically designed for mRNA/gRNA transfection with high efficiency and low toxicity. RmesFect™ is efficient in a large variety of cells.
  • Highly efficient: achieve RNAs delivery in all cells
  • Ready-to-use: no need for additional buffer
  • Low nucleic acid amount: minimized toxicity
  • Protects RNAs against degradation
  • Compatible with any culture medium
  • Serum compatible: Medium changed not required
mRNA transfection provides several advantages over plasmid DNA delivery. No need for nuclear uptake. mRNA translation into proteins occurs in the cytoplasm. It gives faster protein expression than DNA transfection and has no genomic integration. Protein expression in a total promoter-independent manner.