3-Methylhexanoic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methylhexanoicacid

AC45660-0010
103534-300EA 57.08 USD
103534-300 103534-302
3-Methylhexanoic acid 97%
3-Methylhexanoic acid
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 215…216 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00870847
CAS Number: 3780-58-3
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Titration Acid ≥96.0 to ≤104.0 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR