Dibutylboron trifluoromethanesulfonate 1 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC21414-1000
103531-814EA 171.92 USD
103531-814
Dibutylboron trifluoromethanesulfonate 1 M in diethyl ether, AcroSeal™
Dibutylboron trifluoromethanesulfonate
Formula: C₉H₁₈BF₃O₃S
MW: 274.105 g/mol
Density: 0.815 g/cm³
Flash Pt: –40 °C
Storage Temperature: Freezer
MDL Number: MFCD00009669
CAS Number: 60669-69-4
UN: 2924
ADR: 3 I

Order Now

Specification Test Results

Appearance Clear light yellow to orange Solution
Strength 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR