Aluminium nitrate nonahydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum nitrate nonahydrate

AC44634-0025
103531-010EA 104 USD
103531-010 103531-012
Aluminium nitrate nonahydrate ACS
Aluminium nitrate nonahydrate
Formula: Al(NO₃)₃·9H₂O
MW: 375.13 g/mol
Melting Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00149132
CAS Number: 7784-27-2

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.002 %
Sodium (Na) ≤0.005 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR