Monarch® Total RNA Miniprep Enzyme Pack, New England Biolabs
Supplier: New England Biolabs (NEB)
T2019L
103529-162EA 84 USD
103529-162
Monarch® Total RNA Miniprep Enzyme Pack, New England Biolabs
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Need more enzymes for your Monarch® Total RNA Purification Kit? No problem. Monarch® Kit components are all available for purchase separately.
The Monarch® Total RNA Miniprep Enzyme Pack contains lyophilized Monarch® DNase I and Monarch® Proteinase K and their associated reaction buffers and resuspension buffers. The enzymes in this pack are optimized for use with the Monarch® Total RNA Miniprep Kit and are not recommended as substitutes for NEB's standard DNase I and Proteinase K.
Delivery information: The Monarch® Total RNA Miniprep Enzyme pack contains: Monarch® Proteinase K (20.8 mg), Monarch® Proteinase K Resuspension Buffer (1.5 ml), Monarch® Proteinase K Reaction Buffer (4 ml), Monarch® DNase I (275 units), Monarch® DNase I Reaction Buffer (4 ml) and Monarch® Nuclease-free Water (1.5 ml).