Monarch® gDNA Removal Columns, New England Biolabs
Supplier: New England Biolabs (NEB)
T2017L
103529-158EA
174
USD
103529-158
Monarch® gDNA Removal Columns, New England Biolabs
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Monarch gDNA Removal Columns, supplied with the Monarch Total RNA Miniprep Kit, bind genomic DNA during the total RNA miniprep workflow, removing it from your RNA preps. Columns are supplied with Monarch Collection Tubes II , and are compatible with vacuum manifolds.
- A component of the Monarch Total RNA Miniprep Kit
- Binds genomic DNA during the total RNA miniprep workflow to remove it from your RNA prep
- Contain 100 columns and 100 collection tubes
- Also facilitates reduction of sample viscosity as it passes through
- Monarch kit components are also available separately for added convenience
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)