Monarch® RNA Wash Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T2014L
103529-156EA 82 USD
103529-156
Monarch® RNA Wash Buffer, New England Biolabs
Buffers Extraction Buffers
Need more RNA Wash Buffer? No problem. Monarch® kit components are all available for purchase separately.
  • Can be used to wash bound RNA to remove contaminants including salts and enzymes
Monarch® RNA Wash Buffer is a component of the Monarch® Total RNA Miniprep Kit.
Ordering information: Supplied as a 5X concentrate.