Monarch® RNA Priming Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T2013L
103529-154EA 82 USD
103529-154
Monarch® RNA Priming Buffer, New England Biolabs
Buffers Extraction Buffers
Need more RNA Priming Buffer? No problem. Monarch® kit components are all available for purchase separately.
  • Used to wash bound RNA to remove contaminants including salts and enzymes
Monarch® RNA Priming Buffer is a component of the Monarch® Total RNA Miniprep Kit.
Delivery information: Monarch® Priming Buffer is supplied in 2 bottles each containing 28 ml.