Monarch® RNA Lysis Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
T2012L
103529-152EA 82 USD
103529-152
Monarch® RNA Lysis Buffer, New England Biolabs
Buffers Extraction Buffers
Need more RNA Lysis Buffer? No problem. Monarch® Kit components are all available for purchase separately.
  • Can be used to lyse cells and release cellular components, including RNA
The Monarch® RNA Lysis Buffer is a component of the Monarch® Total RNA Miniprep Kit.
Delivery information: Monarch® RNA Lysis Buffer is supplied in two bottles each containing 50 ml.