Monarch® DNA/RNA Protection Reagent, New England Biolabs
Supplier: New England Biolabs (NEB)
T2011L
103529-150EA
129
USD
103529-150
Monarch® DNA/RNA Protection Reagent, New England Biolabs
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
Monarch® DNA/RNA Protection Reagent is a component of the Monarch® Total RNA Miniprep Kit. It is a solution for the preservation of nucleic acids in various tissue types.
- Preserves and stabilizes RNA and DNA
- A component of the Monarch® Total RNA Miniprep Kit (103529-148)
It is supplied as a 2X concentrate. This reagent does not need to be removed before RNA extraction with the Monarch® Total RNA Miniprep Kit.
Delivery information: Supplied as a 2X concentrate. Supplied in two bottles, each containing 28 ml.