Dichloromethane 99+% stabilized, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methane dichloride, Methylene chloride, Methylene chloride

Stabilization: Stabilised with amylene

AC38378-0050
103528-174EA 44.71 USD
103528-174 103528-176 103528-178 103528-180
Dichloromethane 99+% stabilized, extra pure
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39…40 °C (760 mmHg)
Melting Pt: –97 °C
Density: 1.325 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.05 % (K.F.)
GC ≥99.0 %
Stabilizer Approx. 100 ppm Amylene
Residue after evaporation ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR