2,2':6',2''-Terpyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TERPY

AC21563-0050 AC21563-2500
103526-744EA 132 USD
103526-744 103526-746 103526-748
2,2':6',2''-Terpyridine 96%
2,2':6',2''-Terpyridine
Formula: C₁₅H₁₁N₃
MW: 233.27 g/mol
Boiling Pt: 370 °C
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00006213
CAS Number: 1148-79-4
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Appearance Cream to yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 89 to 95 °C
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR