Acrylic acid 98% stabilized, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Prop-2-enoic acid

AC16425-0100
200057-026EA 113 USD
200057-026 200057-024 200057-028 103526-120
Acrylic acid 98% stabilized, Extra Pure
Acrylic acid
Formula: C₃H₄O₂
MW: 72.06 g/mol
Boiling Pt: 139 °C (760 mmHg)
Melting Pt: 13 °C
Density: 1.05 g/cm³
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00004367
CAS Number: 79-10-7
EINECS: 201-177-9
UN: 2218
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR