Chloroacetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

AC14729-0025
103525-638EA 99.4 USD
103525-638 103525-640 103525-642 103525-644
Chloroacetyl chloride 98%
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
MW: 112.94 g/mol
Boiling Pt: 105
Melting Pt: -22.5
Density: 1.42 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1
Merck Index: 13,02074

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR