Dichloroacetic acid (DCA) 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DCA Deblock, DCA

AC11306-2500 AC11306-0010 AC11306-0025
200000-310EA 52.15 USD
200000-310 200000-308 103524-900
Dichloroacetic acid (DCA) 99+%
Dichloroacetic acid (DCA)
Formula: C₂H₂Cl₂O₂
MW: 128.94 g/mol
Boiling Pt: 194
Melting Pt: -4
Density: 1.56 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
EINECS: 201-207-0
UN: 1764
ADR: 8 II
Merck Index: 13,03075

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4650 to 1.4670 (20 °C, 589 nm)
Color scale ≤100 APHA
Titration with NaOH 99.0 to 101.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR