(Trimethylsilyl)diazomethane ∼2 mol/l in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilyldiazomethane, TMS-Diazomethane

AC42920-1000
103524-348EA 567.14 USD
103524-348
(Trimethylsilyl)diazomethane ∼2 mol/l in diethyl ether, AcroSeal™
(Trimethylsilyl)diazomethane
Formula: C₄H₁₀N₂Si
MW: 114.22
Density: 0.77 g/cm³
Flash Pt: –35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053946
CAS Number: 18107-18-1
UN: 1992
ADR: 3,I

Order Now

Specification Test Results

Appearance Yellow Solution (May develop some turbidity or precipitate)
Strength 0.5 % (typically)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR