1,2,3,4-Tetramethylcyclopentadiene mixture of isomers 85%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 1,2,3,4-Tetramethylcyclopenta-1,3-diene, Tetramethylcyclopentadiene

AC34807-0250
200021-546EA 131 USD
200021-546 103524-268
1,2,3,4-Tetramethylcyclopentadiene mixture of isomers 85%
1,2,3,4-Tetramethylcyclopentadiene
Formula: C₉H₁₄
MW: 122.21
Boiling Pt: 142 °C
Density: 0.808 g/cm³
Flash Pt: 42 °C
Storage Temperature: Freezer
MDL Number: MFCD00145385
CAS Number: 4249-10-9
UN: 3295
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4760 (20 °C, 589 nm)
GC ≥80 % (mixture of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR