3'-Azido-3'-deoxythymidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3-Azido-2,3-dideoxy-beta-D-ribofuranosyl)thymine, Zidovudine, Azidothymidine, AZT

AC45498-0010
103517-570EA 42.86 USD
103517-570 103517-572 103517-574
3'-Azido-3'-deoxythymidine 98%
3'-Azido-3'-deoxythymidine
Formula: C₁₀H₁₃N₅O₄
MW: 267.25 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Freezer
MDL Number: MFCD00006536
CAS Number: 30516-87-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 122 to 126 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR