MBD 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 7-(4-Methoxybenzylamino)-4-nitrobenzoxadiazole

AC41354-0050
200006-906EA 159 USD
200006-906 103512-514
MBD 98%
MBD

CAS No.: 33984-50-8

Formula: C₁₄H₁₂N₄O₄
MW: 300.274 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042778
CAS Number: 33984-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR