Adenosine-5'-monophosphoric acid (AMP) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate, 5'-Adenylic acid, Adenosine 5'-monophosphate, Adenosine monophosphate, 5'-AMP

AC10279-1000
200010-982EA 154 USD
200010-982 200005-604 103508-398
Adenosine-5'-monophosphoric acid (AMP) 99%
Adenosine-5'-monophosphoric acid (AMP)
Formula: C₁₀H₁₄N₅O₇P
MW: 347.22 g/mol
Melting Pt: 178…185 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005750
CAS Number: 61-19-8
EINECS: 200-500-0

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤6 % (105 °C, 5 hrs)
HPLC ≥98.5 %
UV spectrophotometry molar absorptivity 15400 +/- 2%, on dry basis
UV transmittance ≥95.0 % (c=1, 0.2 N NaOH, 430 nm) 1 cm cell
Ratio of Absorbances 0.14 to 0.16 (A280/A260)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR