3-Biotin-GTP , New England Biolabs
Supplier: New England Biolabs (NEB)
N0760S
103501-478EA
383
USD
103501-478
3-Biotin-GTP , New England Biolabs
Nucleic Acid Reagents
Nucleotides
Supplied in Ultrapure water as a Triethylammonium salt at pH 7.0
Molecular formula: C₃₁H₅₀N₁₁O₁₉P₃S
Molecular weight: 1005.78 g/mol
Extinction coefficient: 260 = ~11700 Lmol⁻¹ cm⁻¹
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...