NEBNext® Ultra II Directional RNA Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs (NEB)
E7760L E7760S
103501-462EA 3403 USD
103501-462 103501-464
NEBNext® Ultra II Directional RNA Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the 'dUTP' method for strand-specificity, with proven superiority for this application.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE