Anti-RRN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
25918-1-AP-150UL
10345-938EA 462.47 USD
10345-938
Anti-RRN3 Rabbit Polyclonal Antibody
Antibodies
The RRN3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RRN3. This antibody recognizes human, mouse antigen. The RRN3 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: RRN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR