Anti-TCHP Rabbit Polyclonal Antibody

Supplier: Proteintech
25931-1-AP-150UL
10345-906EA 462.47 USD
10345-906
Anti-TCHP Rabbit Polyclonal Antibody
Antibodies
The TCHP antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human TCHP. This antibody recognizes human antigen. The TCHP antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: TCHP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR