Anti-CRHR2 Rabbit Polyclonal Antibody

Supplier: Proteintech
25267-1-AP-150UL
10345-884EA 462.47 USD
10345-884
Anti-CRHR2 Rabbit Polyclonal Antibody
Antibodies
The CRHR2 antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CRHR2. This antibody recognizes human, mouse, rat antigen. The CRHR2 antibody has been validated for the following applications: ELISA, WB analysis.

Type: Primary
Antigen: CRHR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR