Anti-BRCA1 Mouse Monoclonal Antibody (DyLight 680) [clone: RAY]

Supplier: Novus Biologicals
NB100-598FR
103394-336EA 638.77 USD
103394-336
Anti-BRCA1 Mouse Monoclonal Antibody (DyLight 680) [clone: RAY]
Antibodies
The BRCA1 Antibody (RAY) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to BRCA1. This antibody reacts with human. The BRCA1 Antibody (RAY) [DyLight 680] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: RAY
Conjugation: DyLight 680
Epitope: The epitope recognized is found between amino acids 1314-1600.
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR