HiScribe SP6 RNA Synthesis Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E2070S
103391-810EA 319 USD
103391-810
HiScribe SP6 RNA Synthesis Kit, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
The HiScribe SP6 RNA Synthesis Kit is designed for the in vitro transcription of RNA using SP6 RNA Polymerase. This kit is suitable for synthesis of high yield RNA transcripts and for incorporation of cap analogs (not included) or modified nucleotides (not included) to obtain capped, biotin-labeled or dye-labeled RNA.
  • Generate over 80 µg RNA per reaction
  • Enables full substitution of NTPs for labeling and incorporation of modified bases
  • Template removal and mRNA purification reagents included
  • Linearized control template included for verification of RNA synthesis
The kit is also capable of synthesizing high specific activity radiolabeled RNA for use as probes or targets. RNA synthesized from this kit is suitable for many applications including RNA structure and function studies, ribozyme biochemistry, probes for RNase protection or gel shift assays, hybridization-based blots, anti-sense RNA or RNAi experiments, microarray analysis, microinjection, sgRNA synthesis and in vitro translation studies.
This kit contains sufficient reagents for 50 reactions of 25 µl each. Each standard reaction yields ≥80 µg of RNA from 1 µg SP6 Control Template DNA. Each kit can yield ≥4 mg of RNA.