Anti-EPO Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2343R-CY5
10339-030EA 823.27 USD
10339-030
Anti-EPO Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Erythropoietin is the principal hormone involved in the regulation of erythrocyte differentiation and the maintenance of a physiological level of circulating erythrocyte mass.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPO
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR