Human tPA Total Antigens ELISA Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
TPN39-K01
103389-288EA 1484.23 USD
103389-288
Human tPA Total Antigens ELISA Kit, Eagle Biosciences, Inc.
Assays ELISAs
Human Tissue-type Plasminogen Activator Total Antigen ELISA detects total tPA in biological fluids.
  • Dynamic Range: 0.1 - 10 ng/ml
  • Sensitivity: 0.1 ng/ml
  • Incubation: 2 hours
Tissue-type Plasminogen Actvator (tPA) is a member of the serine proteinase family. tPA functions to lyse fibrin clots into soluble plasmin fragments. tPA is active in two forms, single chain and two chain. The two chain tPA is created via interaction with the plasmin product cleaving the single chain. This two chain form is regarded as the more active form.