Myeloperoxidase (MPO) Serum/Plasma ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
MPO31-K01
103389-286EA 876.15 USD
103389-286
Myeloperoxidase (MPO) Serum/Plasma ELISA, Eagle Biosciences
Assays ELISAs
Human Myeloperoxidase (MPO) ELISA is to detect human MPO levels in serum and EDTA-plasma samples.
  • Dynamic Range: 2.0 - 512 ng/ml
  • Sensitivity: 0.65 ng/ml
  • Incubation: 2.5 hours
The Eagle Biosciences Human Myeloperoxidase (MPO) Serum/Plasma ELISA Assay Kit (enzyme-linked immunoassay kit) is intended for the quantitative determination of human myeloperoxidase (MPO) levels in serum and EDTA-plasma samples. The test is useful for detecting elevated levels of myeloperoxidase in serum and plasma samples. The Eagle Biosciences Human Myeloperoxidase (MPO) Serum/Plasma ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.