Antibody to Rituxinab ELISA Assay Kit, Eagle Biosciences
Supplier: Eagle Biosciences
IG-BB106
103389-250EA 2040 USD
103389-250
Antibody to Rituxinab ELISA Assay Kit, Eagle Biosciences
Assays ELISAs
Antibody to Rituximab ELISA to determine free antibodies to Rituximab in serum and plasma samples.
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours
The Eagle Biosciences Antibody to Rituximab is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Rituximab (Rituxan®, Mabthera®) in serum and plasma samples. The Antibody to Rituximab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.