Fecal C. Difficile GDH ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Supplier: Eagle Biosciences
CDF35-K01
103389-160EA 595 USD
103389-160
Fecal C. Difficile GDH ELISA Assay Kit Assay Kit, Eagle Biosciences, Inc.
Assays ELISAs
This Clostridium Difficile GDH ELISA is for the detection of C
  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2 hours
Clostridium difficile is a gram-positive anaerobe. Infection with C.  difficile causes severe diarrhea and can be fatal if not diagnosed and treated in a timely manner. C. difficile infection is induced in patients by long-term treatment with antibiotics and is commonly found in hospital environment.  It is easily transmitted through contact with infected fecal matter. Accurate testing for toxins proved to be difficult due to assays having low sensitivity. Since all strains of C. difficile produce large amounts of glutamate dehydrogenase, testing for this Antigens has proven to be a better screening tool due to its higher negative predictive value.
difficile glutamate dehydrogenase 1.