Mouse / Rat Dopamine ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DOU39-K010
103389-154EA 1281.54 USD
103389-154
Mouse / Rat Dopamine ELISA, Eagle Biosciences
Assays ELISAs
Mouse/Rat Dopamine ELISA detects dopamine in serum, plasma, tissue, and cell culture samples.
  • Dynamic Range: 0.3 - 100 ng/mL
  • Sensitivity: 5.9 pg/mL
  • Incubation: overnight
The Mouse/Rat Dopamine ELISA Assay kit is an Enzyme-linked immunosorbent assay used for the quantitative and very sensitive determination of noradrenaline in biological samples including serum, plasma, tissue, and cell culture samples. The Mouse/Rat Dopamine ELISA Assay kit is for research use only and should not be used in diagnostic procedures.