Mouse / Rat Noradrenaline (Norepinephrine) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
NOU39-K010
103389-150EA 1281.54 USD
103389-150
Mouse / Rat Noradrenaline (Norepinephrine) ELISA, Eagle Biosciences
Assays ELISAs
Mouse/Rat Noradrenaline ELISA detects noradrenaline in biological samples
  • Dynamic Range: 0.05 - 25 ng/mL
  • Sensitivity: 1 pg/mL
  • Incubation: Overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.
For Research Use Only