CIC C3d ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
DCM017
103389-078EA 398.85 USD
103389-078
CIC C3d ELISA, Eagle Biosciences
Assays ELISAs
CIC C3d EIA is colorimetric method to determine Circulating Immune Complex C3d conc in serum,plasma.
  • Dynamic Range: 16 - 64µgEq/ml
  • Sensitivity: 0.60 mgEq/mL
  • Incubation: 1.5 hours
The importance of the immunocomplex (CIC) and their relation with several diseases have been object of investigations for many years.The enstablishment of immunocomplex is a normal protecting process of the immune system. The circulating immunocomplex are removed from the circulation by means of various cellular, biochemical and enzymatic processes.