Ropivacaine hydrochloride ≥98.0% (by HPLC), powder

Supplier: BIOSYNTH INTERNATIONAL INC MS

Synonyms: 2-Piperidinecarboxamide hydrochloride, Ropivacaine HCl

FR65151-250G
103375-122EA 1096.15 USD
103375-122
Ropivacaine hydrochloride ≥98.0% (by HPLC), powder
Ropivacaine hydrochloride
Formula: C₁₇H₂₆N₂O·HCl
MW: 164.64 g/mol
Melting Pt: 260 °C
MDL Number: MFCD06671728
CAS Number: 98717-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR