Anti-DNA Ligase I Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56635
103360-864EA 417.23 USD
103360-864
Anti-DNA Ligase I Rabbit Polyclonal Antibody
Antibodies
The DNA Ligase I Antibody from Novus Biologicals is a rabbit polyclonal antibody to DNA Ligase I. This antibody reacts with human. The DNA Ligase I Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: DNA Ligase I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR