Anti-LOC285697 TAF11 RNA polymerase II Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00285697-B01P
103348-198EA 767.69 USD
103348-198
Anti-LOC285697 TAF11 RNA polymerase II Mouse Polyclonal Antibody
Antibodies
The LOC285697 TAF11 RNA polymerase II Antibody from Novus Biologicals is a mouse polyclonal antibody to LOC285697 TAF11 RNA polymerase II. This antibody reacts with human. The LOC285697 TAF11 RNA polymerase II Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: LOC285697 TAF11 RNA polymerase II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR