Anti-DGK-gamma Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00001608-D01P
103324-858EA 844.62 USD
103324-858
Anti-DGK-gamma Rabbit Polyclonal Antibody
Antibodies
The DGK-gamma Antibody from Novus Biologicals is a rabbit polyclonal antibody to DGK-gamma. This antibody reacts with human. The DGK-gamma Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: DGK-gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR