Anti-UBTF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-2362R-FITC
10332-094EA 823.27 USD
10332-094
Anti-UBTF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recognizes the ribosomal RNA gene promoter and activates transcription mediated by RNA polymerase I through cooperative interactions with the transcription factor SL1/TIF-IB complex. It binds specifically to the upstream control element.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBTF
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR