HistoPro 200 Parafin Infiltration Processor, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62596-10
103307-208EA 7706.57 USD
103307-208
HistoPro 200 Parafin Infiltration Processor, Electron Microscopy Sciences
Tissue Processors
HistoPro® 200 offers unique reagent/paraffin infiltration capability with programmable processing time, temperature, agitation and vacuum in an easy to use, compact, cost conscious package.
  • Up to 100 Cassettes
  • 2 Liter Reagent/Paraffin Capacity
  • Compact footprint
  • Programmable processing time and temperature
  • Programmable vacuum cycle
Certifications: UL listed and CE certified.