Ultra Diamond Knife, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
15-UL 15-ULE 15-ULR 15-US 15-USE 15-USR 18-UL 18-ULE 18-ULR 18-US 18-USE 18-USR 20-ULJ 20-ULJR 20-USJ 20-USJR 21-UL 21-ULE 21-ULR 21-US 21-USE 21-USR 24-UL 24-ULE 24-ULR 24-US 24-USE 24-USR 27-UL 27-ULE 27-ULR 27-US 27-USR 30-UL 30-ULE 30-ULJ 30-ULJR 30-ULR 30-US 30-USE 30-USJ 30-USJR 30-USR 35-UL 35-ULE 35-ULR 35-US 35-USE 35-USR 40-UL 40-ULE 40-ULR 40-US 40-USE 40-USR 30-ULD 35-ULD 40-ULD 15-ULDR 18-ULDR 15-ULDE 21-ULDR 18-ULDE 24-ULDR 21-ULDE 24-ULDE 15-ULD 30-ULDR 30-ULDE 35-ULDR 18-ULD 21-ULD 40-ULDR 35-ULDE 24-ULD 40-ULDE
102680-520EA 5147.42 USD
102680-520 102680-522 102680-524 102680-526 102680-528 102680-530 102680-556 102680-558 102680-560 102680-562 102680-564 102680-566 102680-576 102680-578 102680-580 102680-582 102680-608 102680-610 102680-612 102680-614 102680-616 102680-618 102680-644 102680-646 102680-648 102680-650 102680-652 102680-654 102680-680 102680-682 102680-684 102680-686 102680-690 102680-728 102680-730 102680-732 102680-734 102680-736 102680-744 102680-746 102680-748 102680-750 102680-752 102680-778 102680-780 102680-782 102680-784 102680-786 102680-788 102680-828 102680-830 102680-832 102680-834 102680-836 102680-838 103306-580 103306-586 103306-588 103307-082 103307-108 103307-130 103307-134 103307-142 103307-148 103307-150 103307-152 103307-158 103307-160 103307-162 103307-166 103307-176 103307-178 103307-182 103307-184 103307-194 103307-196 103307-202 103307-204 103307-206
Ultra Diamond Knife, Electron Microscopy Sciences
Knives
These knives are available in 35, and 45 degree angles. The optimal thickness range for cutting with these knives is between 30 and 150 nm. They are available in sizes ranging from 1.5 to 4.0 mm as a standard. All of these knives are tested within a thickness range of 30 to 150 nm.
The 45° recognized as the knife for routine sectioning
The 35° diamond knives have been used for reduced compression, smoother section surfaces and better structural preservation. Also these knives have proved advantageous in Lowicryls, inhomogenous samples such as undecalcified bone, dental materials, sectioning of soft industrial samples such as metals and polymers, as well as for hard and brittle samples, for example semiconductors (Si, GaAs, etc), superconducting oxides, nanocrystalline ceramics.
The ultra 35° knife (in the triangular holder) with a cutting range of 100 to 2 µm is used for dry sectioning of epoxy or acrylic resin embedded biological samples, which need to be investigated by element analysis. The gliding of the sections on the dry knife surface is facilitated with the use of our static line II ionizer.
The ultra diamond knife is now available in Jumbo with 35° and 45° angles and in 2.0 and 3.0 mm sizes.