24-Place SEM Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75958-67
103306-402EA 663.04 USD
103306-402
24-Place SEM Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
This 24 place SEM holder perfect with Hitachi VP SEM.
  • Made of aluminum
  • Unique ID numbers for each hole
This 24-place holder offers a stage travel of 100 x 50 mm. The holder measures 4 7/16 L x 21/2 W x 3/8" H (112.7 x 63.5 x 10 mm) and has a 1/8" (3.2 mm) mounting pin.